Econo™ Lahey Gall Duct Forceps, Floor Grade, Sklar®
Supplier: Sklar
21-453
76192-482BX 186.54 USD
76192-482
Econo™ Lahey Gall Duct Forceps, Floor Grade, Sklar®
Forceps
Econo™ Lahey Gall Duct Forceps are used for blunt dissection and for clamping vessels.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Lahey Gall Duct Forceps have longitudinal serrations on curved tips. The curvature of this instrument allows for dissection in areas that may not be easily accessible. They have ratcheted finger rings for locking into placeMore Product Information