PIPETMAN® Diamond Tips EasyPack™, Gilson®
Supplier: GILSON, INC.
F161451 F161931 F161571 F161210 F161631 F161731 F161671 F161111
76178-148PK 74.06 USD
76178-148 76178-152 76178-180 76178-182 76178-578 76180-020 76180-024 76180-026
PIPETMAN® Diamond Tips EasyPack™, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN pipettes perfectly. They deliver accurate and precise liquid volumes when used with Gilson pipettes. EASYPACK™ is an economical way to order tips in bulk.
  • Accurate and precise
  • Reliable
  • Economical
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy toMore Product Information