Calcium chloride dihydrate ≥99%, white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

0219463505
76177-046EA 582.12 USD
76177-046
Calcium chloride dihydrate ≥99%, white powder cell culture reagent
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176 (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Merck Index: 13,01660
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=012d1dac588a2482dd4270022cdc6f3a0d9cc1ec&vid=c9f5c63a6dd1c97ddfd6fc46a5b392ba3dfeb70b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR