SpectraMax® iD5 Multi-Mode Microplate Reader - Five-Mode Hybrid Microplate Reader with Automatic NFC Filter Identification and Western Blot Capability, Molecular Devices
Supplier: Molecular Devices
ID5-STD 5061514 5056339 0200-7030 6590-0145 6590-0146 6590-0152 ID5-INJ-UPG 6590-0084 6590-0085 6590-0087 6590-0088 6590-0089 6590-0092 6590-0096 6590-0099 6590-0131 6590-0102 6590-0105 6590-0108 6590-0111 6590-0122 6590-0123 6590-0114 6590-0124 6590-0125 6590-0126 6590-0086 6590-0097 6590-0117 6590-0153 6590-0082 6590-0083 6590-0090 6590-0091 6590-0094 6590-0093 6590-0100 6590-0101 6590-0103 6590-0104 6590-0106 6590-0107 6590-0109 6590-0110 6590-0112 6590-0113 6590-0115 6590-0116 6590-0127 6590-0129 6590-0128 6590-0130 6590-0134 6590-0135 6590-0136 6590-0137 6590-0138 6590-0139 6590-0140 6590-0141 6590-0142 6590-0132 6590-0133 6590-0080 6590-0081 6590-0095 6590-0098 6590-0118 6590-0119 6590-0120 6590-0121
76175-288EA 75220.77 USD
76175-288 76175-466 76175-468 76175-470 76175-472 76175-474 76175-476 76175-478 76175-480 76175-482 76175-484 76175-486 76175-488 76175-490 76175-492 76175-494 76175-496 76175-498 76175-500 76175-502 76175-504 76175-506 76175-508 76175-510 76175-512 76175-514 76175-516 76175-518 76175-520 76175-530 76175-532 76175-534 76175-536 76175-538 76175-540 76175-542 76175-544 76175-546 76175-548 76175-550 76175-552 76175-554 76175-556 76175-558 76175-560 76175-562 76175-564 76175-566 76175-568 76175-570 76175-572 76175-574 76175-576 76175-578 76175-580 76175-582 76175-584 76175-592 76175-594 76175-596 76175-598 76175-600 76175-602 76175-604 76175-606 76175-608 76175-610 76175-612 76175-614 76175-616 76175-618 76175-620
SpectraMax® iD5 Multi-Mode Microplate Reader - Five-Mode Hybrid Microplate Reader with Automatic NFC Filter Identification and Western Blot Capability, Molecular Devices
Microplate Readers
The SpectraMax® iD5 Multi-Mode Microplate Reader is the complete laboratory solution to help you increase your research capabilities and comes with built-in absorbance, fluorescence, luminescence, time-resolved fluorescence (TRF), and tunable fluorescence polarization (FP) read modes
  • Easily set up protocols, run experiments, view tutorial videos, and control user access with the intuitive SoftMax Touch Software for the touchscreen
  • Automatically identify filter sets using NFC functionality, simplifying workflows and eliminating confusion
  • Perform
More Product Information