Anti-SLC19A1 IgG Polyclonal Antibody

Supplier: Boster Biological Technology
PB9504
76173-876EA 702.73 USD
76173-876
Anti-SLC19A1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Folate transporter 1(SLC19A1) detection. Tested with WB in Human.

Type: Primary
Antigen: SLC19A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR