VWR® High Performance Multi-Channel Pipettors
Supplier: VWR International
76169-246 76169-248 76169-250 76169-252 76169-254 76169-256 76169-258 76169-260 76208-498 76208-502 76208-504
76169-246EA 824.37 USD
76169-246 76169-248 76169-250 76169-252 76169-254 76169-256 76169-258 76169-260 76208-498 76208-502 76208-504
VWR® High Performance Multi-Channel Pipettors
Pipettes Micropipettes
VWR® High Performance Pipettors are the newest lightweight pipettor designed for user comfort throughout daily work. Thanks to low pipetting forces, users get a unique pipetting experience. With its, high accuracy and precision, the VWR® High Performance Pipettor delivers highly precise and reproducible results.
  • Exclusive volume lock mechanism
  • Lightweight design
  • Low pipetting forces
  • Outstanding precision and accuracy
  • Fully autoclavable without disassembly (120 °C/0.1 MPa/20 min.) and UV resistant
  • Universal tip fitting
  • Unique serial number
Order Now
SPECIFICATIONS
Environmentally Preferable