SpeedDigester IR Digestion System with Suction Module and Condensate Trap, Büchi
Supplier: Buchi
1154251200 1154362200
76169-014EA 8935.07 USD
76169-014 76169-016
SpeedDigester IR Digestion System with Suction Module and Condensate Trap, Büchi
Digestion Systems Mineralization Workstations
Set new standards in terms of digestion speed, flexible use and result reproducibility with the SpeedDigester. Achieve an increased sample throughput for nitrogen determination by Kjeldahl digestion and multiply instrument functionality whenever needs are changing.
  • Two heating chambers for 10 or 12 samples
  • Works with all BUCHI tube sizes (150 mL, 300 mL and 500 mL)
  • Sealed suction module for effective transfer of harmful fumes
  • Increased sample throughput due to shortened process time
  • Individual operation of each heating chamber
  • Fast heat transfer of
More Product Information