Anti-IGF2R Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-6670R-A750
76118-582EA 732.62 USD
76118-582
Anti-IGF2R Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Transport of phosphorylated lysosomal enzymes from the Golgi complex and the cell surface to lysosomes. Lysosomal enzymes bearing phosphomannosyl residues bind specifically to mannose-6-phosphate receptors in the Golgi apparatus and the resulting receptor-ligand complex is transported to an acidic prelyosomal compartment where the low pH mediates the dissociation of the complex. This receptor also binds IGF2. Acts as a positive regulator of T-cell coactivation, by binding DPP4.

Type: Primary
Antigen: IGF2R
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2201-More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR