Anti-Rho GTPase activating protein 29 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-6098R-A680
76117-736EA 732.61 USD
76117-736
Anti-Rho GTPase activating protein 29 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state. Has strong activity toward RHOA, and weaker activity toward RAC1 and CDC42. May act as a specific effector of RAP2A to regulate Rho. In concert with RASIP1, suppresses RhoA signaling and dampens ROCK and MYH9 activities in endothelial cells and plays an essential role in blood vessel tubulogenesis.

Type: Primary
Antigen: Rho GTPase activating protein 29
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/1261
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR