Anti-Proteasome 20S beta 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-9358R-A750
76110-528EA 732.61 USD
76110-528
Anti-Proteasome 20S beta 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This unit is responsible of the peptidyl glutamyl-like activity. May catalyze basal processing of intracellular antigens.

Type: Primary
Antigen: Proteasome 20S beta 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/239
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR