Anti-ASB13 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-9106R-A750
76109-940EA 732.61 USD
76109-940
Anti-ASB13 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the Elongin B/C complex. Several other families of proteins also contain SOCS boxes, but differ from the SOCS proteins in the type of domain they contain upstream of the SOCS box. The largest family of SOCS box-containing proteins is the ankyrin repeat and SOCS box-containing (ASB) protein family. ASB-13 (ankyrin repeat and SOCS box-containing 13) is a 278 amino acid member of the ASB familyMore Product Information