Biometra Thermal Cyclers, Analytik Jena
Supplier: Analytik Jena US
846-4-070-311 846-4-070-301 846-2-070-311 846-2-070-301 846-070-231 846-070-221 846-070-271 846-070-261 846-4-070-280 846-4-070-720 846-4-070-723 846-4-070-724 846-4-070-212 846-4-070-202 846-4-070-213 846-4-070-215 846-070-232 846-070-222 846-070-233 846-070-235 846-4-070-211 846-4-070-201 846-4-070-251 846-4-070-241 846-4-070-210 846-4-070-200 846-4-070-224 846-4-070-214 846-070-230 846-070-220 846-070-234 846-070-224
75873-970EA 9900.7 USD
75873-970 75873-972 75873-974 75873-992 76102-420 76102-422 76102-424 76102-426 76102-428 76102-430 76102-432 76102-434 76102-436 76102-438 76102-440 76102-442 76102-444 76102-446 76102-448 76102-450 76102-486 76102-488 76102-490 76102-492 76102-494 76102-496 76102-498 76102-500 76102-502 76102-504 76102-506 76102-508
Biometra Thermal Cyclers, Analytik Jena
Thermal Cyclers
The Biometra Thermal Cycler product family offers PCR devices ranging from standard to premium equipment – all designed in line with the common principles of being reliable, easy to use, and precise.
  • One block, two block, or 3 block systems available
  • Integrated 7" touchscreen control
  • Whisper quiet, with a low noise emission of 45 dB (max.)
  • High performance smart lid; Defined pressure control for highly reproducible results
  • Linear gradient tool allows easy gradient programming using the primer annealing temperature
  • Heated/Pressurized lid
  • Advanced user
More Product Information