Anti-ATG9A Ser735 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-5199R-A680
76079-922EA 753.8 USD
76079-922
Anti-ATG9A Ser735 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Involved in autophagy and cytoplasm to vacuole transport (Cvt) vesicle formation. Plays a key role in the organization of the preautophagosomal structure/phagophore assembly site (PAS), the nucleating site for formation of the sequestering vesicle. Cycles between a juxta-nuclear trans-Golgi network compartment and late endosomes. Nutrient starvation induces accumulation on autophagosomes. Starvation-dependent trafficking requires ULK1, ATG13 and SUPT20H.

Type: Primary
Antigen: ATG9A Ser735
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR