Anti-Complement C3g fragment Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-4876R-A680
76079-286EA 753.79 USD
76079-286
Anti-Complement C3g fragment Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The complement factor C3 consists of an alpha and a beta chain. C3 is a central factor in the complement cascade. It is central to the alternative pathway that leads to the C3 convertase C3bBb. The classical mannose binding lectin activation pathway leads to the C3 convertase C4b2a. These convertases cleave C3 resulting in C3a and C3b. Further degradation leads to the formation of the alpha chain products C3d, C3g and C3c. C3 is an acute phase protein that is produced by a wide range of tissues, including renal epithelial cells and hepatocytes.

Type: Primary
Antigen: Complement C3gMore Product Information
Order Now

SPECIFICATIONS

Antigen Name Complement C3g fragment
Environmentally Preferable
Antigen Synonyms C3|Complement component 3|C3a anaphylatoxin|Complement C3|Complement C3g fragment|Acylation stimulating protein|Plp|Complement C3 precursor|Complement component C3|ASP|CO3_HUMAN|Complement C3 alpha chain|Complement factor 3
Antibody Type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity
Host Rabbit
Gene ID 718
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 955-1001/1663
Immunogen KLH conjugated synthetic peptide derived from human Complement C3g fragment
Form Liquid
Molecular Weight 5/181 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage Buffer Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
Storage Temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping Temperature 4 °C
Tested Applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR