Anti-USP22 Rabbit Polyclonal Antibody

Supplier: Prosci
61-113
76070-284EA 636.42 USD
76070-284
Anti-USP22 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100

Type: Primary
Antigen: USP22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 351-380
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol USP22
Environmentally Preferable
Antigen Name Ubiquitin carboxyl-terminal hydrolase 22
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 23326
Isotype
Western Blot Yes
ImmunoChemistry Yes
Modification None
Epitope 351-380 aa
Immunogen This USP22 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 351-380 amino acids from the C-terminal region of human USP22.
Form Liquid
Molecular Weight 60 kDa
Purification This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
Concentration 1.17 mg/ml
Storage Temperature Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR