Accredo Safety Small Micro Breaker Lockout, ZING Enterprises
Supplier: ZING Enterprises
7263
76026-364EA 16.15 USD
76026-364
Accredo Safety Small Micro Breaker Lockout, ZING Enterprises
Security Locks
Supports OSHA Standard 1910.147.
  • No Tools Required for Installation
  • High quality
  • Very durable
Small Miniature Circuit Breaker Lockout effectively locks out ISO/DIN miniature circuit breakers. The lockout is constructed from durable aluminum, measures 3/4"H, 1''L, 1/2"W, and accommodates 1 padlock with a maximum shackle diameter of 9/32’’. No tools are required for installation.
Certificaciones: OSHA 1910.147