Samco™ Clicktainer™ Specimen Containers, Thermo Scientific
Supplier: Samco
90WHT53-2000 90WHT53-2002 90WHT53-2003 90WHT53-2001 90WHT53-2004 90WHT53-1002 90WHT53-1001 90ORG53-1000 90GRN53-1000 90ORG53-1001 120WHT53-2000 120ORG53-1000 120WHT53-1000 120WHT53-4000
76019-370CS 155.99 USD
76019-370 76019-374 76019-376 76019-378 76019-380 76019-382 76019-384 76019-386 76019-390 76019-392 76019-396 76019-400 76019-402 76019-408
Samco™ Clicktainer™ Specimen Containers, Thermo Scientific
Sample Containers General Purpose Containers
Unique click seal system provides ultimate sample protection and user safety by ensuring correct application the first time.
  • Visual, audible, and tactile indication of correct cap closure
  • Wide 53 mm opening for ease of sample collection and management
  • Leakproof performance
  • 95 kPa compliant at ambient temperature
  • Ideal for use in pneumatic tube systems
  • Excellent clarity with easy to see graduations
Thermo Scientific understands the value of patient samples.
With this innovative closure system, Clicktainer™ Vials are ideally suited for the collectionMore Product Information