VWR® Beakers without Handle, PP
Supplier: VWR International
76019-260 76019-262 76019-264 76019-266 76019-268 76019-270 76019-272 76019-274 76019-276
76019-260CS 35.22 USD
76019-260 76019-262 76019-264 76019-266 76019-268 76019-270 76019-272 76019-274 76019-276
VWR® Beakers without Handle, PP
Beakers
VWR® polypropylene beakers, which come in a natural color, have a pouring spout and graduations printed directly on the beakers.
  • Autoclavable
  • Integrated pouring lip
  • Stackable
  • Natural color
  • Printed graduation
Since the beakers have no handle, they can be easily stacked to save space. The beakers are autoclavable.
Working temperatures of 0 to 121 °C.
Certificaciones: Resin meets US FDA 21 CFR requirements.
Precaución: Do not stack products while autoclaving.