EZwaste® UN/DOT 70 mm Cap Assembly for HPLC Solvent Waste with Drum Adapter, Foxx Life Sciences
Supplier: VWR International
330-0C03-OEM 330-0C05-OEM 330-0C11-OEM 330-0C12-OEM 330-0C15-OEM 330-0C16-OEM 330-0C18-OEM 330-0C19-OEM 330-0C20-OEM 330-0C21-OEM 330-0C23-OEM
75805-422EA 408.55 USD
75805-422 75805-424 75805-426 75805-428 75805-430 75805-432 76018-624 76018-626 76018-628 76018-630 76018-632 76018-634 76018-636
EZwaste® UN/DOT 70 mm Cap Assembly for HPLC Solvent Waste with Drum Adapter, Foxx Life Sciences
Closures Bottle Closures
This cap system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb options for 1/4" or 3/8" ID
  • Drum adapter included
The EZwaste® UN/DOT Solvent Waste System is designed to be an affordable solution for containing liquidMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable