Cuvette Washer, FireflySci
Supplier: FIREFLYSCI, INC.
P65S P65D P65SKIT P65DKIT P65SKITWPUMP P65DKITWPUMP
76015-784EA 418.05 USD
76015-784 76015-786 76015-788 76015-790 76314-064 76314-066
Cuvette Washer, FireflySci
Glassware Washers
Cuvette washers are handmade and built to last for multiple uses without contamination.
  • Fast cuvette cleaning action
  • Leave cuvettes in place for quick drying action
  • Use with cuvettes from 3 to10mm lightpath
  • Reduces handling and risk of breakage
Easy to use device requires the cuvette to be turned upside down when the cleaning solution is run.
Información para pedidos: Washers includes Gasket and stopper. Vacuum flask and pump must be ordered separately. Double cuvette washer kit includes cuvette washer head, rubber gasket, stopper (size 7),More Product Information