Iodine solution 5% (w/v) USP, strong (DEA List I Chemical)

Supplier: SPECTRUM CHEMICAL MFG CORP

IO106-100ML IO106-200LTBL IO106-20LTBL IO106-4LTGL IO106-4LTGL-CS IO106-500ML-CS IO106-500MLGL
76014-552EA 131.2 USD
76014-552 76014-554 76014-556 76014-558 76014-560 76014-562 76014-564
Iodine solution 5% (w/v) USP, strong (DEA List I Chemical)
Iodine

Strong Iodine Solution, 5 Percent (w/v) (DEA List I Chemical), USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 1.007
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
Merck Index: 13,05036

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR