Shovels, Remco
Supplier: Remco Products
69812 69813 69814 69815 69816
76007-886EA 40.78 USD
76007-886 76007-888 76007-890 76007-892 76007-894
Shovels, Remco
Spades and Shovels
Remco one-piece polypropylene shovels are tough, lightweight and hygienic.
It is a workhorse for moving bulk materials and an integral part of any professional cross–contamination containment, food safety or HACCP program. Initially developed over 30 years ago, our one-piece shovel doesn’t just meet the standard, it sets the standard. This shovel is popular for meat, dairy, seafood, and poultry processing, as well as commercial fishing and other food handling applications.
Certificaciones: FDA compliant raw material.