VWR® Volumetric Flask, Clear Glass, Class A, Unserialized
Supplier: VWR International
76004-016 76004-018 76004-020 76004-022 76004-024 76004-026 76004-028
76004-016CS 207.3 USD
76004-016 76004-018 76004-020 76004-022 76004-024 76004-026 76004-028
VWR® Volumetric Flask, Clear Glass, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certificaciones: Flasks are supplied with Batch certificates.