VWR® Volumetric Flask, Clear Glass, Class A, Serialized
Supplier: VWR International
76004-002 76004-004 76004-006 76004-008 76004-010 76004-012 76004-014
76004-002CS 296.44 USD
76004-002 76004-004 76004-006 76004-008 76004-010 76004-012 76004-014
VWR® Volumetric Flask, Clear Glass, Class A, Serialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certificaciones: Flasks are supplied with individual work certificates.