VWR® Volumetric Flask, Amber, Wide Neck, Class A, Unserialized
Supplier: VWR International
76003-616 76003-618 76003-620 76003-622 76003-624 76003-626 76003-628 76003-630 76003-632 76003-770 76003-772 76003-774 76003-776 76003-786 76003-788 76003-826 76003-828 76003-830 76003-832 76003-834 76003-836 76003-838 76003-840 76003-842 76003-956 76003-958 76003-960 76003-962 76003-964 76004-062 76004-064
76003-616CS 321.78 USD
76003-616 76003-618 76003-620 76003-622 76003-624 76003-626 76003-628 76003-630 76003-632 76003-770 76003-772 76003-774 76003-776 76003-786 76003-788 76003-826 76003-828 76003-830 76003-832 76003-834 76003-836 76003-838 76003-840 76003-842 76003-956 76003-958 76003-960 76003-962 76003-964 76004-062 76004-064
VWR® Volumetric Flask, Amber, Wide Neck, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® volumetric flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certificaciones: Flasks are supplied with Batch certificates.