Chk2 inhibitor 97 HPLC_ASSAY_METHOD

Supplier: Enzo Life Sciences

Synonyms: Chk1 inhibitor II, Chk2 inhibitor I, 5-(2-Amino-5-oxo-1,5-dihydroimidazol-4-ylidine)-3,4,5,10-2H-azepino[3,4-b]indol-1-one

CHM115-0001 CHM115-0500
76002-498EA 513.39 USD
76002-498 76002-974
Chk2 inhibitor 97 HPLC_ASSAY_METHOD
Chk2 inhibitor

Cell-permeable, ATP-competitive, potent inhibitor of Chk2.


Displays anti-proliferative and anti-inflammatory properties. Chk2 inhibitor has also been reported to suppress the growth of leukemic T cells.

Formula: C₁₅H₁₃N₅O₂
MW: 295.3 g/mol
CAS Number: 724708-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d827f07b0dc3c2e43268b249619dd22c8c863605&vid=00ed457916ffb140f5e4fbbabaefa9507e596604&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR