Vista® Basic Large Volume Infusion Pump
Supplier: B BRAUN MEDICAL MS
637-202
75996-452CS 7774.81 USD
75996-452
Vista® Basic Large Volume Infusion Pump
Pumps Syringe Pumps
The Vista basic was designed to ensure simplicity in all aspects of its use: from set installation, to programming, to special mode use and alarm correction. Based on a design that has sold over 100000 units worldwide, the Vista basic provides high reliability packaged in one of the easiest pumps to use.
  • Robust design of mechanical components ensures maximum durability
  • One-year standard warranty and multiple warranty options are available to meet your future service needs
  • Two-year preventive maintenance schedule helps save and money from typical annual
More Product Information