Bioclean™ Polyethylene Pharma Covers, Ansell
Supplier: Ansell Healthcare
BPC-005 BPC-008 BPC-010 BPC-023 BPC-051 BPC-061 BPC-076 BPC-091
75993-436CS 1945.66 USD
75993-436 75993-438 75993-440 75993-442 75993-444 75993-482 75993-486 75993-488 75993-490 75993-492 75993-494
Bioclean™ Polyethylene Pharma Covers, Ansell
Covers
These polyethylene pharma covers provide product protection from environmental impurities during storage, processing and transportation.
  • Breathable microbial barrier
  • Lint-free, moisture and puncture resistant
  • IPA resistant coloured thread
  • Form fitted
  • Fast drying time
  • Easy to apply and remove
  • Autoclavable at 121 to 127 °C
Pharma Covers minimize the need for autoclave tape during wrapping and save time when prepping glassware and while tubing for autoclaving. Easy removal minimizes particulate generation and facilitates aseptic technique.