HardyDisks™ Levofloxacin, LVX-5, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8611 Z8615
75993-464EA 24.37 USD
75993-464 75993-466
HardyDisks™ Levofloxacin, LVX-5, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.