PCR Tubes, Watson Bio Lab
Supplier: Watson Bio Lab
137-211C 137-212C 137-231C 137-232C 137-333C 137-431C 137-432C
75982-952CS 2945.53 USD
75982-952 75982-954 75982-956 75982-958 75982-960 75982-962 75982-964
PCR Tubes, Watson Bio Lab
Tubes PCR Tubes
The side walls are uniformly thin, which enables PCR with superior heat exchange.
  • Thin wall, high transparency
  • Nuclease-free certified
Flat and dome caps are available.
All tubes feature autoclavable polypropylene construction.
The flat top, smooth and superior in optical transparency, is the most suitable for Real Time PCR. They also offer writing space.
Order Now
SPECIFICATIONS
Environmentally Preferable