BioClean™ Bouffant Caps, BBC, Ansell
Supplier: Ansell Healthcare
BBC-W
75972-518CS 377.83 USD
75972-518
BioClean™ Bouffant Caps, BBC, Ansell
Disposable Garments
The BioClean™ bouffant cap is lightweight and breathable and reduces the risk of contamination from the head area from entering the controlled environment.
  • Used in life sciences industry
  • Latex-free elastic
  • Stitched seams
  • Weight: 10 GSM
Soft bouffant caps are made from spunbond polypropylene with elasticated double stitching.
Envase: Caps are packaged 100 per sealed inner PE bag; 10 inner bags per sealed outer PE bag; one outer bag per lined carton (1000 pieces).