TRIO.BAS™ Base Station Induction Battery Charger
Supplier: Hardy Diagnostics
BAS318
75928-106EA 2146.82 USD
75928-106
TRIO.BAS™ Base Station Induction Battery Charger
Samplers Air Samplers
Induction battery charging base station for use with the TRIO.BAS™ MONO, DUO, TRIO, and Isolator air sampling units.
  • 100/240 VCA
  • 50/60 Hz
  • 35 W US variant plug
Units simply rest on top of the base station and begin charging.
Order Now
SPECIFICATIONS
Environmentally Preferable