Vimentin IHC Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
3870A 3870B
75876-930BX 91.27 USD
75876-930 75876-932
Vimentin IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Vimentin control slides contain sections of positive staining melanoma and negative staining adipose.
  • Tissue type: (+) Tonsil / (-) Adipose
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide
Newcomer supply vimentin control slides are for the positive immunohistochemical staining of Vimentin, a ubiquitous intermediate filament foundMore Product Information