Eppendorf® Vacufuge Plus Concentrator
Supplier: Eppendorf
022822101 022820109 022820001 022820168 022822993
53512-636EA 94.78 USD
53512-636 47745-390 53512-622 53512-624 53512-620 75875-186 75875-188
Eppendorf® Vacufuge Plus Concentrator
Concentrators
The eppendorf vacufuge plus facilitates fast and efficient, gentle vacuum concentration of DNA, RNA, nucleotides, proteins and other liquid or wet samples with its advanced heating technology. Its coated lid provides superior chemical resistance against many acids and organic solvents.
  • Comes in 3 system configurations
  • Extremely quiet operation <50 dB(A)
  • Chemical-resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
  • Small footprint saves valuable bench space, Coated lid with superior chemical resistance
  • Wide variety of
More Product Information