SP Scientific miVac Swing Rotors and Components, Genevac
Supplier: Genevac
DRS-00000-200 QRS-00000-400 BRS-STK00-004 BRS-STK00-016
75871-596EA 1882.97 USD
75871-596 75871-598 75871-600 75871-602
SP Scientific miVac Swing Rotors and Components, Genevac
Centrifuges Rotors
Configure your miVac Concentrator for use with microtitre shallow- or deep-well plates.
  • For use with deep-well or shallow-well plates
  • Stackers increase capacity of rotors
  • Durable stainless steel
miVac Swing rotors allow for use of microtitre plates in concentrators and can be combined with stackers for additional capacity.
Use these rotors with deep-well or shallow-well plates. Optional sets of 4 stackers increase capacity from 2 or 4 plates to 8 plates and can be used with all miVac concentrators. The set of 16 stackers increases capacity from 4More Product Information