CORIO™ Refrigerated/Heating Circulators
Supplier: JULABO
9012703.02 9012704.02 9012701.02 9012707.02 9012707.13 9970337 9012727.13 9012713.N1.02 9012713.N1.13 9012713.S1.02 9012713.S1.13 9012714.N1.02 9012714.N1.13 9012714.S1.02 9012714.S1.13 9012727.02 9012701.N1.02 9012701.N1.13 9012715.N1.02 9012715.N1.13 9012717.N1.02 9012717.N1.13 9012728.N1.02 9012728.N1.13
10015-750EA 7437.64 USD
10015-750 10015-752 10015-754 10015-818 75839-420 75839-422 75839-424 75839-566 75871-446 75871-448 75871-450 75871-452 76558-262 76558-234 76558-166 76558-212 76558-216 76558-272 76558-268 76558-190 76558-240 76558-200 77442-060 77442-062 77442-064 77442-066 77442-068 77442-070 77442-072 77442-074
CORIO™ Refrigerated/Heating Circulators
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
CORIO™ refrigerated/heating circulators for routing liquid temperature control applications from –40 to +150 °C. A great value with German engineering and quality.
  • Stainless steel construction
  • Adjustable pump
  • German quality
  • USB communication
  • Quiet operation
The new generation of economy CORIO™ refrigerated/heating circulators from JULABO provide fast and reliable temperature control for internal or external applications.
The units incorporate a USB port for connection to an external PC. This enables control via PLC or the free JULABO EasyTEMPMore Product Information