VWR® Storage Carts
Supplier: VWR International
10809-048 10809-050 10809-052 10809-054 10809-056 75857-058 75857-060 75857-062 75857-066 75857-070 75857-072 75857-074 75857-076 75857-078 75857-080 75857-084 75857-086 75857-088 75857-090 75857-092 75857-094 75857-098 75857-100 75857-102 75857-104 75857-106 75857-108 75857-110 75857-112 75857-118 75857-122 75857-124 75857-126 75857-128 75857-130 75857-132 75857-134 75870-342 75870-344 75870-350 75870-352 75870-360 75870-364 75870-366 75870-374 75870-378 75870-380 75870-382 75870-384 75870-388 75870-390 75870-394 75870-400
10809-048EA 2124.41 USD
10809-048 10809-050 10809-052 10809-054 10809-056 75857-058 75857-060 75857-062 75857-066 75857-070 75857-072 75857-074 75857-076 75857-078 75857-080 75857-084 75857-086 75857-088 75857-090 75857-092 75857-094 75857-098 75857-100 75857-102 75857-104 75857-106 75857-108 75857-110 75857-112 75857-118 75857-122 75857-124 75857-126 75857-128 75857-130 75857-132 75857-134 75870-342 75870-344 75870-350 75870-352 75870-360 75870-364 75870-366 75870-374 75870-378 75870-380 75870-382 75870-384 75870-388 75870-390 75870-394 75870-400
VWR® Storage Carts
Carts
Storage carts made from food-grade, high-density polyethylene have four 3" industrial casters, two of which lock.
  • Locking hinged door
  • Bulk storage area with door
  • One key locks drawers and bulk storage
  • Full-extension drawers with handle
  • Polyethylene construction resists chemicals
The carts feature full-extension drawers for full viewing of drawer contents.
The carts add extra storage and work space in labs, clinics, hospitals, and other locations. They move all supplies together in a single work area, so no one needs to make extra trips to set up anMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable