MF Series Mechanical Force Gauges, SEALS USA INC
Supplier: Nidec Shimpo America
MF-2 MF-5 MF-10 MF-20 MF-30 MF-50 MF-100 MF-1KG MF-2KG MF-3KG MF-5KG MF-10KG MF-20KG MF-30KG MF-50KG
75854-112EA 821.26 USD
75854-112 75854-114 75854-116 75854-118 75854-120 75854-122 75854-124 75854-126 75854-128 75854-130 75854-132 75854-134 75854-136 75854-138 75854-158
MF Series Mechanical Force Gauges, SEALS USA INC
Dynamometers
MF Series Mechanical Force Gauge is excellent for push/pull force testing in manufacturing, material testing, maintenance, inspection applications
  • Outstanding ±0.2% F.S. accuracy makes the MF’s Ideal even for the strictest applications
  • Precision gauge movement provides a quick response for instantaneous analysis
  • Measures Tension and Compression Forces – Only one gauge required for insertion withdrawal tests
  • Real Time Mode or Peak Mode enables flexibility to view live readings or maximum values recorded
  • Smooth mechanical operation with precise action
More Product Information