Forced Air Fine Oven, Yamato
Supplier: Yamato Scientific
DF-832 DF-1032 DH-832 DH-1032
75846-410EA 13095.53 USD
75846-410 75846-412 75846-678 75846-680
Forced Air Fine Oven, Yamato
Ovens Standard Ovens
This oven has high-accuracy temperature control and a manual exhaust damper.
  • Forced Air Circulation and Ventilation System
  • PID Z Control Temperature Controller
  • Stainless Steel Pipe Heater with Controllable Fan
  • Double K Thermocouple Sensor
  • Cable Port, 30 mm dia.
The interior is made of stainless steel, and an exhaust damper allows quick exhaust and cooling of the inside chamber
Self-diagnostic functions, a key lock function, a door switch, fan failure detection, and an electric leakage breaker with over-current protection are provided. The oven alsoMore Product Information