Standard Laboratory Use Sterilizer, Yamato
Supplier: Yamato Scientific
SN-510C SN-300C SN-500C
97003-658EA 11811.94 USD
97003-658 75846-576 75846-580
Standard Laboratory Use Sterilizer, Yamato
Sterilizers Autoclaves (Steam Sterilizers)
Features of the ergonomically designed, easy-to-use sterilizer include easy top loading and sample handling.
  • Wheels for Mobility Provided
  • Two Heaters
  • Lid Interlock Mechanism
  • Three Female Thread Ports for Sensors
  • Axial Fan Motor
The maximum sterilizing temperature goes up to 135 °C, and the operating temperature range is 45 to 135 °C. With simple setting and operation, this sterilizer manages ordinary sterilization, sterilization of culture mediums and liquids, and melting of culture mediums with its automatic high-pressure steam sterilization system.
More Product Information