Forced Convection Temperature Oven, Yamato
Supplier: Yamato Scientific
DKN-302C DKN-312C DKN-402C DKN-412C DKN-602C DKN-612C DKN-812C DKN-912C
75846-340EA 3521.09 USD
75846-340 75846-398 75846-400 75846-402 75846-522 75846-524 75846-526 75846-528
Forced Convection Temperature Oven, Yamato
Ovens Standard Ovens
Standard forced air convection ovens come with extended functions and safety features.
  • Forced Air Circulation Operating Method
  • Programmable PID Controller
  • Built-in Exhaust Ports and Scirocco Fans
  • Cable Hole: 30 mm dia. on Right Side
They feature exact temperature accuracy and are easy to program
The ovens also feature safety functions such as self-diagnostic abilities, a key lock function, and automatic overheating prevention. The interior is made of stainless steel, and the exterior is made of a cold-steel plate with melamine-resin baking finish.More Product Information