Full Color Polarizing Polariscope with Swivel Mount, Ace Glass
Supplier: Ace Glass
14400-10
75846-508EA 1503.44 USD
75846-508
Full Color Polarizing Polariscope with Swivel Mount, Ace Glass
Polarimeters
Full color polarizer cabinet for viewing strain patterns in glass and other transparent materials.
  • Swivel mount allows cabinet to be locked into any horizontal or vertical position
Polariscope consists of a 4" deep cabinet with a 10x19" illuminated area, and a 5" square viewer.
Información para pedidos: One-year manufacturer’s warranty.