Temperature Controllers, Ramp and Soak Profile, 1200 Watt, Ace Glass
Supplier: Ace Glass
12333-35 12333-15 12333-20 12333-25 12333-30 12333-10
75846-448EA 3667.14 USD
75846-448 75846-480 75846-482 75846-484 75846-486 75846-748
Temperature Controllers, Ramp and Soak Profile, 1200 Watt, Ace Glass
Controllers Temperature Controllers
Single outlet, ramp and soak stepping temperature controller audible alarm and 1200 watts of output
  • LED is user selectable for Fahrenheit or Centigrade
Temperature, Time, Hold, Soak and End steps are programmed via the PC EZ-ZONE software and RS-485 interface cable (sold separately). Four profiles of ten steps each, you can link profiles for a total of 40 steps.
Información para pedidos: Includes integral rack mounting bracket. One-year limited manufacturer's warranty. Download available for the the EZ-ZONE configuration software, data loggingMore Product Information