Programmable Natural Convection Oven, Yamato
Supplier: Yamato Scientific
DVS-402C DVS-412C DVS-602C DVS-612C
75846-332EA 3194.6 USD
75846-332 75846-334 75846-336 75846-338
Programmable Natural Convection Oven, Yamato
Ovens Standard Ovens
These constant-temperature ovens feature easy-to-use program settings.
  • Six-Pattern PID Program Controller
  • Simultaneous Display of Set Constant and Measured Temperature
  • Cable Hole: 30 mm dia. on Right Side
  • Exhaust Port: Two 30 mm dia. on Top
  • Two Stainless Steel Plates with Four Brackets Provided
They take approximately 90 minutes (room temperature: ±5 to 260 °C) to heat
The interior material is stainless steel, and the exterior is made of a cold-rolled steel plate with melamine resin baking finish. The glass observation window is chemicallyMore Product Information