Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RPA-T4]

Supplier: BIOGEMS INTERNATIONAL INC.
06121-77-100
75844-012EA 309.69 USD
75844-012
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RPA-T4]
Antibodies
The RPA-T4 monoclonal antibody specifically binds to the CD4 receptor for the human immunodeficiency virus (HIV). CD4 is a 59 kDa single-chain transmembrane glycoprotein that expressed on the surface of most of the thymocytes, T-helper cells, and in low levels on monocytes and macrophages. CD4 is a co-receptor in the antigen-induced T cell activation, together with the MHC class II. The RPA-T4 antibody is capable of blocking HIV binding and inhibiting syncytium formation, by binding to the D1 domain of the CD4 antigen. The OKT4 and the RPA-T4 monoclonal antibodies recognize differentMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol CD4
Environmentally Preferable
Antigen Synonyms Leu-3|L3T4|T4
Antibody Type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)-Cy7®
Clone RPA-T4
Reactivity Human
Host Mouse
Isotype IgG1 kappa
Flow Cytometry Yes
Form liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR