Anti-TER-119 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TER-119]

Supplier: BIOGEMS INTERNATIONAL INC.
81212-60-25
75843-554EA 77.42 USD
75843-554
Anti-TER-119 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TER-119]
Antibodies
The TER-119 monoclonal antibody specifically binds to the mouse 52 kDa Glycophorin A expressed on the erythroid lineage cells, from the early proerythroblast to mature erythrocyte stages, in the fetal liver, adult bone marrow, newborn liver, yolk sac, and adult lymphoid organs. The TER-119 antibody is used, together with other antibodies, as a marker for hematopoietic lineages cells.

Type: Primary
Antigen: TER-119
Clonality: Monoclonal
Clone: TER-119
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR