Anti-IFNG Mouse Monoclonal Antibody [clone: NIB42]

Supplier: BIOGEMS INTERNATIONAL INC.
80821-25-50
75843-494EA 205.56 USD
75843-494
Anti-IFNG Mouse Monoclonal Antibody [clone: NIB42]
Antibodies
The NIB42 monoclonal antibody specifically reacts with human interferon gamma, a multifunctional cytokine that is produced mainly by activated T and NK cells. Interferon gamma has anti-viral and anti-parasitic properties and plays an important role in the activation, growth, and differentiation of macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-α, to inhibit proliferation of normal and transformed cells. The NIB42 antibody neutralizes IFN-γ.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: NIB42
Conjugation:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR