Rat IgG1 Isotype Control (PE (Phycoerythrin)) [clone: HRPN]

Supplier: BIOGEMS INTERNATIONAL INC.
42813-60-100
75842-474EA 161.3 USD
75842-474
Rat IgG1 Isotype Control (PE (Phycoerythrin)) [clone: HRPN]
Antibodies
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR