Rat IgG2b Isotype Control (APC (Allophycocyanin)-Cy7®) [clone: LTF-2]

Supplier: BIOGEMS INTERNATIONAL INC.
41823-87-100
75842-464EA 212.91 USD
75842-464
Rat IgG2b Isotype Control (APC (Allophycocyanin)-Cy7®) [clone: LTF-2]
Antibodies
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR