HardyDisk™ AST , Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8361 Z9121 Z8851 Z9011 Z9241 Z9125 Z9015 Z8855 Z8241 Z8245 Z8311 Z8315 Z8275 Z8905 Z8321 Z8325 Z8411 Z8575 Z8365 Z9245 Z8735 Z8791 Z8795 Z8815
10753-968EA 34.23 USD
10753-968 10753-970 10753-972 10753-974 10753-976 75832-484 75832-486 75832-488 75832-490 75832-492 75832-494 75832-496 75832-498 75832-502 75832-504 75832-506 75832-508 75832-512 75832-516 75832-722 75832-724 75832-726 75832-728 75832-732
HardyDisk™ AST , Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST’s are impregnated paper disks used for antimicrobial susceptibility testing (AST), also known as disk diffusion or Kirby-Bauer testing
Fully compatible with the BBL single or multiple disk dispensers.
Información para pedidos: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.